Product Details

SNP ID
rs111562988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:810870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGAAGTTTAATACCAGTGAGCT[A/G]CGTAAGATTGCGTTAAAAAAAAAAA
Phenotype
MIM: 605247 MIM: 609059 MIM: 180530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIDD1 PubMed Links

Gene Details

Gene
PIDD1
Gene Name
p53-induced death domain protein 1
There are no transcripts associated with this gene.

Gene
PNPLA2
Gene Name
patatin like phospholipase domain containing 2
There are no transcripts associated with this gene.

Gene
RPLP2
Gene Name
ribosomal protein lateral stalk subunit P2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004.3 Intron NP_000995.1
Gene
SNORA52
Gene Name
small nucleolar RNA, H/ACA box 52
There are no transcripts associated with this gene.

View Full Product Details