Product Details

SNP ID
rs114656588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8518257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAATGCATCAAAGAGAAATCAG[A/G]ACTGAAAAGTGCTGAAGGTACAGAG
Phenotype
MIM: 609964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLEC4D PubMed Links

Gene Details

Gene
CLEC4D
Gene Name
C-type lectin domain family 4 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080387.4 408 Missense Mutation GAA,GGA E72G NP_525126.2
XM_011520632.2 408 Missense Mutation GAA,GGA E72G XP_011518934.1
XM_011520633.2 408 Missense Mutation GAA,GGA E72G XP_011518935.1

View Full Product Details