Product Details

SNP ID
rs112643478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8689356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAAAGAGACGTCCTCGTTGGGA[A/G]CAGCCTAGTTTTGCTTTGTTGCCAA
Phenotype
MIM: 614054
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIMKLB PubMed Links

Gene Details

Gene
RIMKLB
Gene Name
ribosomal modification protein rimK like family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297776.1 Intron NP_001284705.1
NM_020734.3 Intron NP_065785.2
XM_006719118.2 Intron XP_006719181.1
XM_006719127.2 Intron XP_006719190.1
XM_011520776.1 Intron XP_011519078.1
XM_011520777.1 Intron XP_011519079.1
XM_011520778.2 Intron XP_011519080.1
XM_011520779.2 Intron XP_011519081.1
XM_011520780.1 Intron XP_011519082.1
XM_011520783.1 Intron XP_011519085.1
XM_011520790.1 Intron XP_011519092.1
XM_017019677.1 Intron XP_016875166.1
XM_017019678.1 Intron XP_016875167.1
XM_017019679.1 Intron XP_016875168.1
XM_017019680.1 Intron XP_016875169.1
XM_017019681.1 Intron XP_016875170.1
XM_017019682.1 Intron XP_016875171.1
XM_017019683.1 Intron XP_016875172.1
XM_017019684.1 Intron XP_016875173.1
XM_017019685.1 Intron XP_016875174.1
XM_017019686.1 Intron XP_016875175.1
XM_017019687.1 Intron XP_016875176.1
XM_017019688.1 Intron XP_016875177.1
XM_017019689.1 Intron XP_016875178.1
XM_017019690.1 Intron XP_016875179.1
XM_017019691.1 Intron XP_016875180.1
XM_017019692.1 Intron XP_016875181.1
XM_017019693.1 Intron XP_016875182.1
XM_017019694.1 Intron XP_016875183.1
XM_017019695.1 Intron XP_016875184.1
XM_017019696.1 Intron XP_016875185.1
XM_017019697.1 Intron XP_016875186.1

View Full Product Details