Product Details

SNP ID
rs114597208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:24994327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAATTGTCATAGCATGATGTAG[C/G]CTGATTTAAAAAAAGAAGAAAAAAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C12orf77 PubMed Links

Gene Details

Gene
C12orf77
Gene Name
chromosome 12 open reading frame 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101339.2 624 Missense Mutation AGC,AGG S136R NP_001094809.2
XM_017018966.1 624 Missense Mutation AGC,AGG S208R XP_016874455.1
Gene
LOC645177
Gene Name
uncharacterized LOC645177
There are no transcripts associated with this gene.

View Full Product Details