Product Details

SNP ID
rs114949843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7479969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACACAGAAATTAGTTCAGCAGCA[C/T]TGAAATCAGCTGACTCATGGGAATT
Phenotype
MIM: 605545
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD163 PubMed Links

Gene Details

Gene
CD163
Gene Name
CD163 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004244.5 3393 Missense Mutation AAT,AGT N1124S NP_004235.4
NM_203416.3 3393 Intron NP_981961.2
XM_005253528.4 3393 Intron XP_005253585.1
XM_005253529.4 3393 Missense Mutation AAT,AGT N1092S XP_005253586.1
XM_017020231.1 3393 Intron XP_016875720.1

View Full Product Details