Product Details
- SNP ID
-
rs117653207
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:5437687 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAACGAGGCAGGCAGTTTCTGGC[G/T]TTGGGGATGTTGAACTAATGACCGT
- Phenotype
-
MIM: 162660
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NTF3
PubMed Links
Gene Details
- Gene
- NTF3
- Gene Name
- neurotrophin 3
View Full Product Details