Product Details

SNP ID
rs116577757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6621054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGGTGAAGAGCAGGTCGCTGGC[C/T]GCCAGGTTACACATGTACACGCTCA
Phenotype
MIM: 606926
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LPAR5 PubMed Links

Gene Details

Gene
LPAR5
Gene Name
lysophosphatidic acid receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142961.1 476 Silent Mutation NP_001136433.1
NM_020400.5 476 Silent Mutation NP_065133.1

View Full Product Details