Product Details

SNP ID
rs111766137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6905937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGCTGGCTCATGCTTATGTCAA[A/G]CTGGAGGTTAAAGAGAGGTGCGTTT
Phenotype
MIM: 131360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENO2 PubMed Links

Gene Details

Gene
ENO2
Gene Name
enolase 2
There are no transcripts associated with this gene.

Gene
LRRC23
Gene Name
leucine rich repeat containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135217.1 439 Silent Mutation AAA,AAG K73K NP_001128689.1
NM_006992.3 439 Silent Mutation AAA,AAG K73K NP_008923.1
NM_201650.2 439 Silent Mutation AAA,AAG K73K NP_964013.1

View Full Product Details