Product Details

SNP ID
rs116572599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110381161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGGTGACAAATCCAGTGAAAGC[C/T]CTAAATAAATGTAAAATGCCTAATG
Phenotype
MIM: 606949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANAPC7 PubMed Links

Gene Details

Gene
ANAPC7
Gene Name
anaphase promoting complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137664.1 Intron NP_001131136.1
NM_016238.2 Intron NP_057322.2
XM_005253903.3 Intron XP_005253960.1
XM_011538454.2 Intron XP_011536756.1
XM_017019425.1 Intron XP_016874914.1
XM_017019426.1 Intron XP_016874915.1
XM_017019427.1 Intron XP_016874916.1
XM_017019428.1 Intron XP_016874917.1

View Full Product Details