Product Details

SNP ID
rs114172135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112141017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGGGGAGGAAAAGAGAAATGAG[A/G]TTGCCATACCTCCTAATGCATATGA
Phenotype
MIM: 613197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAFD1 PubMed Links

Gene Details

Gene
TRAFD1
Gene Name
TRAF-type zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143906.1 522 Missense Mutation ATT,GTT I146V NP_001137378.1
NM_006700.2 522 Missense Mutation ATT,GTT I146V NP_006691.1

View Full Product Details