Product Details

SNP ID
rs112291500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99982472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCGGCGCCGGGCGGCGGGCAGCA[C/G]GGGCTGTTCGGGCCGGGCGCGGGCG
Phenotype
MIM: 603073
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZIC2 PubMed Links

Gene Details

Gene
ZIC2
Gene Name
Zic family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007129.3 472 Missense Mutation CAC,CAG H136Q NP_009060.2
XM_011521110.2 472 Missense Mutation CAC,CAG H136Q XP_011519412.1

View Full Product Details