Product Details

SNP ID
rs117757664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:101458503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCTTGATTAAACAGCATATAA[A/G]AGCTATTAAGAGTCCAAATATAGGA
Phenotype
MIM: 604234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGBL1 PubMed Links

Gene Details

Gene
ITGBL1
Gene Name
integrin subunit beta like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271754.1 Intron NP_001258683.1
NM_001271755.1 Intron NP_001258684.1
NM_001271756.1 Intron NP_001258685.1
NM_004791.2 Intron NP_004782.1
XM_005254100.4 Intron XP_005254157.1

View Full Product Details