Product Details

SNP ID
rs111957036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1436044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTAGCAGCTGCACGGATGATAG[C/T]AGCGTGGCCAGCTCCTGCCGCAGGG
Phenotype
MIM: 602727
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC154 PubMed Links

Gene Details

Gene
CCDC154
Gene Name
coiled-coil domain containing 154
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143980.1 1697 Silent Mutation CTA,CTG L510L NP_001137452.1
Gene
CLCN7
Gene Name
chloride voltage-gated channel 7
There are no transcripts associated with this gene.

Gene
LOC105371045
Gene Name
uncharacterized LOC105371045
There are no transcripts associated with this gene.

View Full Product Details