Product Details

SNP ID
rs118203998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23603471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTACCCTAACTTATGAATAGTG[G/C]TATACAAATATATTTCCATCTTTTT
Phenotype
MIM: 603836 MIM: 610355
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
NDUFAB1 PubMed Links

Gene Details

Gene
NDUFAB1
Gene Name
NADH:ubiquinone oxidoreductase subunit AB1
There are no transcripts associated with this gene.

Gene
PALB2
Gene Name
partner and localizer of BRCA2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024675.3 4282 Nonsense Mutation TAC,TAG Y1183* NP_078951.2
XM_011545946.2 4282 Nonsense Mutation TAC,TAG Y1185* XP_011544248.1
XM_011545947.2 4282 UTR 3 XP_011544249.1
XM_011545948.2 4282 Nonsense Mutation TAC,TAG Y888* XP_011544250.1
XM_017023671.1 4282 Nonsense Mutation TAC,TAG Y1106* XP_016879160.1
XM_017023672.1 4282 Nonsense Mutation TAC,TAG Y1104* XP_016879161.1
XM_017023673.1 4282 UTR 3 XP_016879162.1

View Full Product Details