Product Details

SNP ID
rs113803159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28887457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTGCAGCCGGCAAGGCCTTGGG[C/G]ATCGTGGCCACCACTGGTGTGGGCA
Phenotype
MIM: 108730
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP2A1 PubMed Links

Gene Details

Gene
ATP2A1
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286075.1 847 Silent Mutation GGC,GGG G96G NP_001273004.1
NM_004320.4 847 Silent Mutation GGC,GGG G221G NP_004311.1
NM_173201.3 847 Silent Mutation GGC,GGG G221G NP_775293.1
Gene
ATP2A1-AS1
Gene Name
ATP2A1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details