Product Details

SNP ID
rs113627401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2971237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGCAAGGATGGCTGGCTGGGA[A/G]GCACACATTGCGTGGCCTGCCTTGC
Phenotype
MIM: 609899 MIM: 614578 MIM: 602474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KREMEN2 PubMed Links

Gene Details

Gene
KREMEN2
Gene Name
kringle containing transmembrane protein 2
There are no transcripts associated with this gene.

Gene
PAQR4
Gene Name
progestin and adipoQ receptor family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284511.1 677 Intron NP_001271440.1
NM_001284512.1 677 Intron NP_001271441.1
NM_001284513.1 677 Missense Mutation AGC,GGC S16G NP_001271442.1
NM_001324118.1 677 Missense Mutation AGC,GGC S83G NP_001311047.1
NM_152341.4 677 Missense Mutation AGC,GGC S83G NP_689554.2
Gene
PKMYT1
Gene Name
protein kinase, membrane associated tyrosine/threonine 1
There are no transcripts associated with this gene.

View Full Product Details