Product Details

SNP ID
rs112242011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29816743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGGCGCAGGGAGAAGTCCCCA[A/G]CGCTGGGGGAGAAGAGCCTGCCGAG
Phenotype
MIM: 600999 MIM: 605088 MIM: 612033 MIM: 614386
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAZ PubMed Links

Gene Details

Gene
MAZ
Gene Name
MYC associated zinc finger protein
There are no transcripts associated with this gene.

Gene
MVP
Gene Name
major vault protein
There are no transcripts associated with this gene.

Gene
PAGR1
Gene Name
PAXIP1 associated glutamate rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024516.3 537 Missense Mutation AAC,AGC N73S NP_078792.1
Gene
PRRT2
Gene Name
proline rich transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256442.1 537 Intron NP_001243371.1
NM_001256443.1 537 Intron NP_001243372.1
NM_145239.2 537 Intron NP_660282.2
XM_011545715.2 537 Intron XP_011544017.1
XM_011545716.2 537 Intron XP_011544018.1
XM_017022887.1 537 Intron XP_016878376.1
XM_017022888.1 537 Intron XP_016878377.1
XM_017022889.1 537 Intron XP_016878378.1

View Full Product Details