Product Details

SNP ID
rs112691371
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30856895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAATTCCTAAGAATATGTCAGAG[C/T]TTTGCTCTCTATTGGATATGGCATG
Phenotype
MIM: 605847
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
BCL7C PubMed Links

Gene Details

Gene
BCL7C
Gene Name
BCL tumor suppressor 7C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286526.1 Intron NP_001273455.1
NM_004765.3 Intron NP_004756.2
XM_011545980.2 Intron XP_011544282.1
XM_017023885.1 Intron XP_016879374.1
XM_017023886.1 Intron XP_016879375.1
XM_017023887.1 Intron XP_016879376.1
XM_017023888.1 Intron XP_016879377.1

View Full Product Details