Product Details
- SNP ID
-
rs114810024
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:760783 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAGGCTGCCAGGCTCTCCACCCC[C/T]ACTTCCCAATTGAGGAAACCGAGGC
- Phenotype
-
MIM: 601051
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR662
PubMed Links
Gene Details
- Gene
- MIR662
- Gene Name
- microRNA 662
There are no transcripts associated with this gene.
- Gene
- MSLN
- Gene Name
- mesothelin
View Full Product Details