Product Details

SNP ID
rs114761341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57512278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCCCCACAACCCCCGCCCACAT[C/T]TGCCATACTTTCAGATGCCCCAAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC102A PubMed Links

Gene Details

Gene
CCDC102A
Gene Name
coiled-coil domain containing 102A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033212.3 2329 UTR 3 NP_149989.2
XM_011523469.2 2329 UTR 3 XP_011521771.1

View Full Product Details