Product Details

SNP ID
rs114602408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69435206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGGAGATAATAGCACATACTT[A/C]ACAGGGTAGTTGTGGTTGTGACAAG
Phenotype
MIM: 611964
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CYB5B PubMed Links

Gene Details

Gene
CYB5B
Gene Name
cytochrome b5 type B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030579.2 Intron NP_085056.2

View Full Product Details