Product Details

SNP ID
rs115754951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70690941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTCCAAATGTGCATTTACTGAC[C/T]AAGTAACCAAGAACCCAGAGAGGGC
Phenotype
MIM: 616951 MIM: 604632
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTSS1L PubMed Links

Gene Details

Gene
MTSS1L
Gene Name
metastasis suppressor 1 like
There are no transcripts associated with this gene.

Gene
VAC14
Gene Name
Vac14, PIKFYVE complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018052.3 3564 Intron NP_060522.3
XM_005256038.3 3564 UTR 3 XP_005256095.1
XM_005256041.3 3564 Intron XP_005256098.2
XM_011523223.2 3564 Intron XP_011521525.1
XM_011523224.2 3564 Intron XP_011521526.1
XM_011523225.2 3564 Intron XP_011521527.1
XM_017023441.1 3564 Intron XP_016878930.1
XM_017023442.1 3564 Intron XP_016878931.1

View Full Product Details