Product Details

SNP ID
rs118106739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71475217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCTGGGAGGCCAAAACGACAAA[C/T]GTCCAGCACAGGCTTCTCTCCAGAG
Phenotype
MIM: 194525
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF19 PubMed Links

Gene Details

Gene
ZNF19
Gene Name
zinc finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006961.3 1599 Missense Mutation ATT,GTT I444V NP_008892.2

View Full Product Details