Product Details

SNP ID
rs114362471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75298960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACCTTCAATTGATTGATCAGAC[A/G]GCAGCAGCTGGGACTAGCCAGTGTT
Phenotype
MIM: 608108
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFDP1 PubMed Links

Gene Details

Gene
CFDP1
Gene Name
craniofacial development protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006324.2 Intron NP_006315.1
XM_011522814.1 Intron XP_011521116.1
XM_011522815.1 Intron XP_011521117.1

View Full Product Details