Product Details

SNP ID
rs111685508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84191766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTTGGTCCCTATGCTCAGAGAC[A/G]CCGCCGGAGCAGGACCTGGCCTGAG
Phenotype
MIM: 604905
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAD2 PubMed Links

Gene Details

Gene
ADAD2
Gene Name
adenosine deaminase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145400.1 111 Intron NP_001138872.1
NM_139174.3 111 Intron NP_631913.3
XM_005255814.1 111 Intron XP_005255871.1
XM_011522919.1 111 Intron XP_011521221.1
XM_011522920.1 111 UTR 5 XP_011521222.1
Gene
TAF1C
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit C
There are no transcripts associated with this gene.

View Full Product Details