Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001159377.1 | 2793 | UTR 3 | NP_001152849.1 | ||
NM_001159378.1 | 2793 | UTR 3 | NP_001152850.1 | ||
NM_001159379.1 | 2793 | UTR 3 | NP_001152851.1 | ||
NM_001159380.1 | 2793 | UTR 3 | NP_001152852.1 | ||
NM_022764.2 | 2793 | UTR 3 | NP_073601.2 | ||
XM_005256101.1 | 2793 | UTR 3 | XP_005256158.1 | ||
XM_005256105.1 | 2793 | UTR 3 | XP_005256162.1 | ||
XM_005256106.1 | 2793 | UTR 3 | XP_005256163.1 | ||
XM_006721247.3 | 2793 | UTR 3 | XP_006721310.1 | ||
XM_011523280.2 | 2793 | UTR 3 | XP_011521582.1 | ||
XM_011523282.2 | 2793 | Intron | XP_011521584.1 | ||
XM_011523283.2 | 2793 | Intron | XP_011521585.1 | ||
XM_011523284.2 | 2793 | Intron | XP_011521586.1 | ||
XM_011523285.1 | 2793 | UTR 3 | XP_011521587.1 | ||
XM_011523286.1 | 2793 | UTR 3 | XP_011521588.1 | ||
XM_011523287.1 | 2793 | UTR 3 | XP_011521589.1 | ||
XM_011523288.1 | 2793 | UTR 3 | XP_011521590.1 | ||
XM_011523289.1 | 2793 | UTR 3 | XP_011521591.1 | ||
XM_017023571.1 | 2793 | UTR 3 | XP_016879060.1 | ||
XM_017023572.1 | 2793 | UTR 3 | XP_016879061.1 | ||
XM_017023573.1 | 2793 | UTR 3 | XP_016879062.1 | ||
XM_017023574.1 | 2793 | UTR 3 | XP_016879063.1 |