Product Details

SNP ID
rs116055738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88573490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCTTTTGTTGCCTTGGGCTGCA[A/G]AAATGCTCTGTGCATCAAGCGCCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZC3H18 PubMed Links

Gene Details

Gene
ZC3H18
Gene Name
zinc finger CCCH-type containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294340.1 Intron NP_001281269.1
NM_144604.3 Intron NP_653205.3
XM_011522863.1 Intron XP_011521165.1
XM_011522865.1 Intron XP_011521167.1
XM_017022934.1 Intron XP_016878423.1
Gene
ZC3H18-AS1
Gene Name
ZC3H18 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details