Product Details

SNP ID
rs117505012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89213501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGTTAAAGTACTTCCAAAATTA[A/G]CTTTAAAATACCACGCACAGGCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF778 PubMed Links

Gene Details

Gene
ZNF778
Gene Name
zinc finger protein 778
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201407.1 Intron NP_001188336.1
NM_182531.3 Intron NP_872337.2
XM_005256288.2 Intron XP_005256345.1
XM_011522940.2 Intron XP_011521242.1
XM_017023015.1 Intron XP_016878504.1
XM_017023016.1 Intron XP_016878505.1
XM_017023017.1 Intron XP_016878506.1

View Full Product Details