Product Details

SNP ID
rs116222098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89270191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCTCATCTGTGCCCACATCAGC[A/C]CCTTTCTGAGAGCTGCAGTATGACA
Phenotype
MIM: 611192
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD11 PubMed Links

Gene Details

Gene
ANKRD11
Gene Name
ankyrin repeat domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256182.1 8651 Intron NP_001243111.1
NM_001256183.1 8651 Intron NP_001243112.1
NM_013275.5 8651 Intron NP_037407.4
XM_006721181.1 8651 Intron XP_006721244.1
XM_006721184.2 8651 Intron XP_006721247.1
XM_011523051.2 8651 Intron XP_011521353.1
XM_011523053.2 8651 Intron XP_011521355.1
XM_011523054.2 8651 Intron XP_011521356.1
XM_011523055.2 8651 Intron XP_011521357.1
XM_011523056.2 8651 Intron XP_011521358.1
XM_011523057.2 8651 UTR 3 XP_011521359.1
XM_017023182.1 8651 Intron XP_016878671.1
XM_017023183.1 8651 Intron XP_016878672.1
XM_017023184.1 8651 Intron XP_016878673.1
XM_017023185.1 8651 Intron XP_016878674.1
XM_017023186.1 8651 Intron XP_016878675.1
XM_017023187.1 8651 Intron XP_016878676.1

View Full Product Details