Product Details

SNP ID
rs114722904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:12672533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCCAATTTTTGTTAAAATAAAA[C/T]GTATCATTTATCCCCAGAAATTTAG
Phenotype
MIM: 606127
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928418 PubMed Links

Gene Details

Gene
LOC101928418
Gene Name
uncharacterized LOC101928418
There are no transcripts associated with this gene.

Gene
MYOCD
Gene Name
myocardin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146312.2 Intron NP_001139784.1
NM_153604.3 Intron NP_705832.1
XM_005256863.1 Intron XP_005256920.1
XM_005256864.1 Intron XP_005256921.1
XM_017025342.1 Intron XP_016880831.1

View Full Product Details