Product Details

SNP ID
rs116262834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17843937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCACAGATAGGAAGGGCTGGTGA[C/T]GGGCACTGGGGGAGGCGAGCACTTC
Phenotype
MIM: 184756 MIM: 615519
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SREBF1 PubMed Links

Gene Details

Gene
SREBF1
Gene Name
sterol regulatory element binding transcription factor 1
There are no transcripts associated with this gene.

Gene
TOM1L2
Gene Name
target of myb1 like 2 membrane trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033551.2 5108 UTR 3 NP_001028723.1
NM_001082968.1 5108 UTR 3 NP_001076437.1
NM_001288786.1 5108 UTR 3 NP_001275715.1
NM_001288787.1 5108 UTR 3 NP_001275716.1
NM_001288788.1 5108 UTR 3 NP_001275717.1
NM_001288789.1 5108 UTR 3 NP_001275718.1
XM_005256461.2 5108 UTR 3 XP_005256518.1
XM_005256462.1 5108 UTR 3 XP_005256519.1
XM_005256463.2 5108 UTR 3 XP_005256520.1
XM_005256466.1 5108 UTR 3 XP_005256523.1
XM_011523662.1 5108 UTR 3 XP_011521964.1
XM_017024201.1 5108 UTR 3 XP_016879690.1

View Full Product Details