Product Details

SNP ID
rs114237735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:745069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCACAGACCTCCGGGTGGAGCA[C/T]GGCCATGATGTGCAGAGCATGGCAG
Phenotype
MIM: 611627 MIM: 606969
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DBIL5P PubMed Links

Gene Details

Gene
DBIL5P
Gene Name
diazepam binding inhibitor-like 5, pseudogene
There are no transcripts associated with this gene.

Gene
FAM57A
Gene Name
family with sequence similarity 57 member A
There are no transcripts associated with this gene.

Gene
GEMIN4
Gene Name
gem nuclear organelle associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015721.2 3311 Missense Mutation ATG,GTG M992V NP_056536.2
XM_005256667.4 3311 Missense Mutation ATG,GTG M996V XP_005256724.1
XM_005256670.4 3311 Missense Mutation ATG,GTG M981V XP_005256727.1
XM_011523910.2 3311 Missense Mutation ATG,GTG M996V XP_011522212.1
XM_011523911.2 3311 Missense Mutation ATG,GTG M996V XP_011522213.1
XM_011523912.2 3311 Missense Mutation ATG,GTG M981V XP_011522214.1
XM_011523913.2 3311 Missense Mutation ATG,GTG M981V XP_011522215.1
XM_017024709.1 3311 Missense Mutation ATG,GTG M996V XP_016880198.1

View Full Product Details