Product Details
- SNP ID
-
rs114933901
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:4989911 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTACCAGCATGGGTGGAATGTGCTG[C/T]TCCTCCAGCCAAGTGGGGCTGTGAA
- Phenotype
-
MIM: 611508
MIM: 603060
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAMTA2
PubMed Links
Gene Details
- Gene
- CAMTA2
- Gene Name
- calmodulin binding transcription activator 2
There are no transcripts associated with this gene.
- Gene
- INCA1
- Gene Name
- inhibitor of CDK, cyclin A1 interacting protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001167985.1 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
NP_001161457.1 |
NM_001167986.1 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
NP_001161458.1 |
NM_001167987.1 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
NP_001161459.1 |
NM_213726.2 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
NP_998891.2 |
XM_005256627.3 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
XP_005256684.1 |
XM_005256628.4 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
XP_005256685.1 |
XM_005256629.1 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
XP_005256686.1 |
XM_006721517.1 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
XP_006721580.1 |
XM_006721518.2 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
XP_006721581.1 |
XM_006721519.3 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
XP_006721582.1 |
XM_011523832.2 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
XP_011522134.1 |
XM_011523834.2 |
653 |
Silent Mutation |
GAA,GAG |
E59E |
XP_011522136.1 |
- Gene
- KIF1C
- Gene Name
- kinesin family member 1C
There are no transcripts associated with this gene.
- Gene
- LOC101927979
- Gene Name
- translation initiation factor IF-2
There are no transcripts associated with this gene.
View Full Product Details