Product Details

SNP ID
rs111897235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7688823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCGGTTGTCCCCAGATCCTGTG[A/G]CTGGCTCAGCTGTGTCCCAGGAGCT
Phenotype
MIM: 191170 MIM: 612661
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TP53 PubMed Links

Gene Details

Gene
TP53
Gene Name
tumor protein p53
There are no transcripts associated with this gene.

Gene
WRAP53
Gene Name
WD repeat containing antisense to TP53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143990.1 330 Missense Mutation ACT,GCT T59A NP_001137462.1
NM_001143991.1 330 Missense Mutation ACT,GCT T59A NP_001137463.1
NM_001143992.1 330 Missense Mutation ACT,GCT T59A NP_001137464.1
NM_018081.2 330 Missense Mutation ACT,GCT T59A NP_060551.2
XM_011523952.2 330 UTR 5 XP_011522254.1
XM_017024812.1 330 Missense Mutation ACT,GCT T59A XP_016880301.1

View Full Product Details