Product Details

SNP ID
rs117175349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8173191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTACTGATGTGCAATGTTTCCTG[A/G]GAAGTGAGGATTAAAGTTATCAAAC
Phenotype
MIM: 616663 MIM: 616183
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNORD118 PubMed Links

Gene Details

Gene
SNORD118
Gene Name
small nucleolar RNA, C/D box 118
There are no transcripts associated with this gene.

Gene
TMEM107
Gene Name
transmembrane protein 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032354.3 1545 UTR 3 NP_115730.2
NM_183065.2 1545 UTR 3 NP_898888.1

View Full Product Details