Product Details
- SNP ID
-
rs113849507
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:50561554 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACCTGTCGGGGGCCGGGGGCCGGC[C/G]GGGGCCGGGGTCAGCAGAAAAGGAC
- Phenotype
-
MIM: 604065
MIM: 613939
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CACNA1G
PubMed Links
Gene Details
- Gene
- CACNA1G
- Gene Name
- calcium voltage-gated channel subunit alpha1 G
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256324.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243253.1 |
NM_001256325.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243254.1 |
NM_001256326.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243255.1 |
NM_001256327.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243256.1 |
NM_001256328.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243257.1 |
NM_001256329.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243258.1 |
NM_001256330.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243259.1 |
NM_001256331.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243260.1 |
NM_001256332.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243261.1 |
NM_001256333.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243262.1 |
NM_001256334.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243263.1 |
NM_001256359.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243288.1 |
NM_001256360.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243289.1 |
NM_001256361.1 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_001243290.1 |
NM_018896.4 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_061496.2 |
NM_198376.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938190.1 |
NM_198377.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938191.2 |
NM_198378.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938192.1 |
NM_198379.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938193.1 |
NM_198380.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938194.1 |
NM_198382.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938196.1 |
NM_198383.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938197.1 |
NM_198384.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938198.1 |
NM_198385.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938199.1 |
NM_198386.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938200.1 |
NM_198387.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938201.1 |
NM_198388.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938202.1 |
NM_198396.2 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
NP_938406.1 |
XM_006722160.3 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
XP_006722223.1 |
XM_006722161.3 |
487 |
Missense Mutation |
CCG,CGG |
P32R |
XP_006722224.1 |
- Gene
- CACNA1G-AS1
- Gene Name
- CACNA1G antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- SPATA20
- Gene Name
- spermatogenesis associated 20
There are no transcripts associated with this gene.
View Full Product Details