Product Details

SNP ID
rs117043883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59700492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGCATAGATGACAGCCCTGGAA[A/G]TACTTATAAATTTCCTTTATGTTCT
Phenotype
MIM: 118955 MIM: 608625 MIM: 611753
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLTC PubMed Links

Gene Details

Gene
CLTC
Gene Name
clathrin heavy chain
There are no transcripts associated with this gene.

Gene
PTRH2
Gene Name
peptidyl-tRNA hydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015509.2 Intron NP_001015509.1
NM_016077.4 Intron NP_057161.1
XM_011524887.1 Intron XP_011523189.1
Gene
VMP1
Gene Name
vacuole membrane protein 1
There are no transcripts associated with this gene.

View Full Product Details