Product Details
- SNP ID
-
rs117742403
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:59718081 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTACCTGCTGATCCATTATTGGGA[A/G]ATTAATGGAGATTCTGAATGTTTTT
- Phenotype
-
MIM: 611753
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VMP1
PubMed Links
Gene Details
- Gene
- VMP1
- Gene Name
- vacuole membrane protein 1
There are no transcripts associated with this gene.
View Full Product Details