Product Details

SNP ID
rs113595573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:66966295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTTAAGCGTGGCGCTCAGATC[A/G]GGGACCGGGTGGAGGGAGTTGGTGG
Phenotype
MIM: 606404
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNG4 PubMed Links

Gene Details

Gene
CACNG4
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014405.3 Intron NP_055220.1

View Full Product Details