Product Details

SNP ID
rs114327289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74331262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTCTGATGGGGCCAGCTGGCC[C/T]GTCAAAGCTCTGTCCTTCCTCCTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF19 PubMed Links

Gene Details

Gene
KIF19
Gene Name
kinesin family member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153209.3 Intron NP_694941.2
XM_005257029.1 Intron XP_005257086.1
XM_005257030.1 Intron XP_005257087.1
XM_017024150.1 Intron XP_016879639.1
XM_017024151.1 Intron XP_016879640.1
XM_017024152.1 Intron XP_016879641.1
XM_017024153.1 Intron XP_016879642.1
XM_017024154.1 Intron XP_016879643.1
XM_017024155.1 Intron XP_016879644.1
XM_017024156.1 Intron XP_016879645.1
XM_017024157.1 Intron XP_016879646.1
XM_017024158.1 Intron XP_016879647.1

View Full Product Details