Product Details
- SNP ID
-
rs111582384
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:79024858 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTCACTGTTCTCTGGTGCCTTTC[G/T]GAGGGTGTGGAAGGAAGAGGAATGG
- Phenotype
-
MIM: 610365
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C1QTNF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199958974,rs538441999] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1QTNF1
- Gene Name
- C1q and tumor necrosis factor related protein 1
- Gene
- C1QTNF1-AS1
- Gene Name
- C1QTNF1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details