Product Details

SNP ID
rs111744243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81120545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTGCCTGGGCCCCCCATCCGCT[G/T]TTGTGGGGCCGGCCCTGACAACAGT
Phenotype
MIM: 605276 MIM: 605475 MIM: 614059
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AATK PubMed Links

Gene Details

Gene
AATK
Gene Name
apoptosis associated tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080395.2 3316 Missense Mutation NP_001073864.2
NM_004920.2 3316 Missense Mutation NP_004911.2
XM_006722193.2 3316 Missense Mutation XP_006722256.1
XM_006722194.2 3316 Missense Mutation XP_006722257.1
XM_006722195.2 3316 Missense Mutation XP_006722258.1
XM_006722196.2 3316 Missense Mutation XP_006722259.1
XM_011525509.2 3316 Missense Mutation XP_011523811.1
XM_011525510.2 3316 Missense Mutation XP_011523812.1
XM_011525511.2 3316 Missense Mutation XP_011523813.1
XM_017025421.1 3316 Missense Mutation XP_016880910.1
XM_017025422.1 3316 Missense Mutation XP_016880911.1
Gene
BAIAP2
Gene Name
BAI1 associated protein 2
There are no transcripts associated with this gene.

Gene
MIR3065
Gene Name
microRNA 3065
There are no transcripts associated with this gene.

Gene
MIR338
Gene Name
microRNA 338
There are no transcripts associated with this gene.

Gene
MIR657
Gene Name
microRNA 657
There are no transcripts associated with this gene.

View Full Product Details