Product Details

SNP ID
rs117656401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81308216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCAGCACCCCCTAGCTCACTGC[C/T]TGGCAACCCCACTCCTGCTGGTGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00482 PubMed Links

Gene Details

Gene
LINC00482
Gene Name
long intergenic non-protein coding RNA 482
There are no transcripts associated with this gene.

Gene
TMEM105
Gene Name
transmembrane protein 105
There are no transcripts associated with this gene.

View Full Product Details