Product Details

SNP ID
rs112188737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42781139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTAAGGGTCCCTGAAGCTGTGGC[C/G]CTAGAGCGGCGGCGGGAGCAGGAAG
Phenotype
MIM: 610907 MIM: 601844
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
VPS25 PubMed Links

Gene Details

Gene
VPS25
Gene Name
vacuolar protein sorting 25 homolog
There are no transcripts associated with this gene.

Gene
WNK4
Gene Name
WNK lysine deficient protein kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321299.1 509 UTR 5 NP_001308228.1
NM_032387.4 509 Silent Mutation GCC,GCG A147A NP_115763.2
XM_011525135.2 509 Silent Mutation GCC,GCG A147A XP_011523437.1
XM_017024962.1 509 Silent Mutation GCC,GCG A147A XP_016880451.1
XM_017024963.1 509 Silent Mutation GCC,GCG A147A XP_016880452.1
XM_017024964.1 509 Silent Mutation GCC,GCG A147A XP_016880453.1
XM_017024965.1 509 Silent Mutation GCC,GCG A147A XP_016880454.1
XM_017024966.1 509 Intron XP_016880455.1

View Full Product Details