Product Details

SNP ID
rs113722041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44926998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCCAAACCTCACCTCGCAACG[C/T]GCTTCTTCTTGCAGGCAGAGGTCCC
Phenotype
MIM: 614570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF18B PubMed Links

Gene Details

Gene
KIF18B
Gene Name
kinesin family member 18B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001264573.1 2526 Missense Mutation CAC,CGC H798R NP_001251503.1
NM_001265577.1 2526 Missense Mutation CAC,CGC H786R NP_001252506.1
XM_011524385.2 2526 Missense Mutation CAC,CGC H807R XP_011522687.1
XM_011524386.2 2526 Missense Mutation CAC,CGC H798R XP_011522688.1
XM_011524387.2 2526 Missense Mutation CAC,CGC H798R XP_011522689.1
XM_011524388.2 2526 Missense Mutation CAC,CGC H795R XP_011522690.1
XM_011524389.2 2526 Missense Mutation CAC,CGC H807R XP_011522691.1
XM_011524390.2 2526 Missense Mutation CAC,CGC H772R XP_011522692.1
XM_011524391.2 2526 Intron XP_011522693.1
Gene
MIR6783
Gene Name
microRNA 6783
There are no transcripts associated with this gene.

View Full Product Details