Product Details

SNP ID
rs115923887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35549778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTACAGATTCCATGATTTTACA[C/T]ATGCAGTTCCTAGGGCCTTTACTCC
Phenotype
MIM: 614958
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLFN14 PubMed Links

Gene Details

Gene
SLFN14
Gene Name
schlafen family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129820.1 Intron NP_001123292.1
XM_017024576.1 Intron XP_016880065.1
XM_017024577.1 Intron XP_016880066.1
XM_017024578.1 Intron XP_016880067.1
XM_017024579.1 Intron XP_016880068.1

View Full Product Details