Product Details

SNP ID
rs114664198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35745226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCTTCCGGAGAGTTCTCCTGCC[C/T]TTGCTCAGACATGCCTTGGCTTTGT
Phenotype
MIM: 611398 MIM: 612128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAS2L2 PubMed Links

Gene Details

Gene
GAS2L2
Gene Name
growth arrest specific 2 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139285.3 2299 Silent Mutation AAA,AAG K757K NP_644814.1
Gene
RASL10B
Gene Name
RAS like family 10 member B
There are no transcripts associated with this gene.

View Full Product Details