Product Details

SNP ID
rs111614697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35998362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGGATGCTTTGTGAGATGTAGG[A/G]GGTGCAGCAGTCAGCAGCAAAGTGA
Phenotype
MIM: 601393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL15 PubMed Links

Gene Details

Gene
CCL15
Gene Name
C-C motif chemokine ligand 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032965.5 712 Missense Mutation CCC,TCC P56S NP_116741.2
Gene
CCL15-CCL14
Gene Name
CCL15-CCL14 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
LOC101927339
Gene Name
uncharacterized LOC101927339
There are no transcripts associated with this gene.

View Full Product Details