Product Details

SNP ID
rs116346098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12794024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTTGACATGTATGAATACAGT[C/T]GCAAAATTTACCAGTTTTTAACAAA
Phenotype
MIM: 176887
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPN2 PubMed Links

Gene Details

Gene
PTPN2
Gene Name
protein tyrosine phosphatase, non-receptor type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207013.1 1763 Intron NP_001193942.1
NM_001308287.1 1763 UTR 3 NP_001295216.1
NM_002828.3 1763 UTR 3 NP_002819.2
NM_080422.2 1763 Intron NP_536347.1
NM_080423.2 1763 Intron NP_536348.1
XM_005258124.3 1763 UTR 3 XP_005258181.1
XM_005258125.3 1763 Intron XP_005258182.1
XM_011525705.2 1763 UTR 3 XP_011524007.1
XM_011525706.2 1763 UTR 3 XP_011524008.1
XM_011525707.2 1763 UTR 3 XP_011524009.1
XM_017025884.1 1763 Intron XP_016881373.1
XM_017025885.1 1763 UTR 3 XP_016881374.1
XM_017025886.1 1763 UTR 3 XP_016881375.1
XM_017025887.1 1763 Intron XP_016881376.1
XM_017025888.1 1763 Intron XP_016881377.1

View Full Product Details