Product Details

SNP ID
rs116865748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35334157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTTCTCGGTAACACTGTGAAAG[A/G]AAATATCCTTTTTCATTTTACATTT
Phenotype
MIM: 194534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF24 PubMed Links

Gene Details

Gene
ZNF24
Gene Name
zinc finger protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308123.1 4360 Intron NP_001295052.1
NM_006965.3 4360 UTR 3 NP_008896.2
XM_005258341.3 4360 UTR 3 XP_005258398.1

View Full Product Details